Learn More
Nitritex™ Polyurethane Tacky Mat
Keep dirt, dust and bacteria at bay in controlled laboratory environments with the Nitritex™ Polyurethane Tacky Mat.
Brand:  Nitritex™ STW1845
87.58 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15258318
  • 143.00€ / Pack of 4
Estimated Shipment: 23-06-2025
Description
  • Matting for use in controlled laboratory environments
  • Rectangular
  • Polyurethane
  • White or blue
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 9001:2015
30 Sheets/Mat, 4 Mats/Cs.
Polyurethane
18 x 45 in.
White
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nitritex™ Polyurethane Tacky Mat > Size: 18 x 45in., White