Learn More
Cytiva Polyvinyl Chloride Fume Cupboard
Product Code. p-8009240
Click to view available options
Width (Metric):
120 cm
150 cm
180 cm
Unit Size:
Each
Product Code. 12596596
Brand: Cytiva 2642
This rich, robust, reliable, fume cupboard made of polyvinylchloride features glare fluorescent tube lightening.
  • Fume cupboard made of polyvinylchloride with glare fluorescent tube lightening
  • Standard sorbridge PVC (= registry fume cupboards)
  • PVC rear panel over the entire width of the fume hood
  • Guide the air at the top and bottom
  • Uses all types of solvents, volatile products, powders, condensing
Specifications
Product Type Fume Cupboard
Lighting Type Glare Fluorescent Tube
Material PVC
Certifications/Compliance NFX 15-210 class B & EN 14175
Quantity 1 Ea.
Width (Metric) 150 cm