Learn More
Haldenwanger™ Porcelain Buchner Funnel
Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 127C/4A
 View more versions of this product
Product Code. 10027440
  • 266.00€ / Each
Estimated Shipment: 17-06-2025
Description
  • Available in a variety of sizes
  • Available with perforated fixed filter plate
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
1250 mL
235 mm
For Filter Paper
1Each
Porcelain
155 mm
155 mm
Buchner Funnel
Documents
One moment while we fetch your results.
Product Title
Haldenwanger™ Porcelain Buchner Funnel > Diameter Plate: 155mm; Capacity: 1.25mL