Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
Brand:  Rogo-Sampaic PNF014
 View more versions of this product
Product Code. 11747335
  • 194.00€ / Each
Only null left
Estimated Shipment: 02-07-2025
Description
  • White porcelain
  • Available for a range of volumes and filter diameters
  • Applications include collecting recrystalized compounds
  • Excellent thermal and chemical resistance
Specifications
Specifications
1080 mL
150mm
96mm
Buchner Funnel
Porcelain
150mm
1Each
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Rogo-Sampaic Porcelain Buchner Funnels > Diameter Plate: 150mm; Capacity: 1080mL