Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
Brand:  Rogo-Sampaic PNF009
 View more versions of this product
Product Code. 11797325
  • 58.00€ / Each
Only null left
Estimated Shipment: 07-07-2025
Add to basket
Description
Description
  • White porcelain
  • Available for a range of volumes and filter diameters
  • Applications include collecting recrystalized compounds
  • Excellent thermal and chemical resistance
Specifications
Specifications
80 mL
55mm
64mm
Buchner Funnel
Porcelain
55mm
1Each
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Rogo-Sampaic Porcelain Buchner Funnels > Diameter Plate: 55mm; Capacity: 80mL