Learn More
Fisherbrand™ Porcelain Plate For Desiccators
For use with desiccators
Brand:  Fisherbrand™ 9180003210
 View more versions of this product
Product Code. 15504655
  • 135.00€ / Each
Estimated Shipment: 15-05-2025
Description
  • For use with desiccators
Specifications
Specifications
280 mm
Plate
Desiccator (Dia.: 300mm)
Certificates
View accessory products
Product Title
Fisherbrand™ Porcelain Plate For Desiccators > Plate diameter: 280mm; Desiccator diameter: 300mm