missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Porcine Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
643.00€ - 4040.00€
Specifications
Species | Porcine |
---|---|
Binding Target | Multiplex Cytokine Assay Kits |
Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742394
|
R&D Systems
LXSAPM-15 |
15 Plex |
3285.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18751954
|
R&D Systems
LXSAPM-14 |
14 Plex |
3100.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732024
|
R&D Systems
LXSAPM-09 |
9 Plex |
2160.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732464
|
R&D Systems
LXSAPM-04 |
4 Plex |
1135.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742424
|
R&D Systems
LXSAPM-05 |
5 Plex |
1340.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732354
|
R&D Systems
LXSAPM-17 |
17 Plex |
3655.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762284
|
R&D Systems
LXSAPM-13 |
13 Plex |
2920.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792074
|
R&D Systems
LXSAPM-06 |
6 Plex |
1545.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722354
|
R&D Systems
LXSAPM-16 |
16 Plex |
3470.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732164
|
R&D Systems
LXSAPM-10 |
10 Plex |
2370.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722154
|
R&D Systems
LXSAPM-12 |
12 Plex |
2735.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752054
|
R&D Systems
LXSAPM-18 |
18 Plex |
3835.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722304
|
R&D Systems
LXSAPM-03 |
3 Plex |
929.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762224
|
R&D Systems
LXSAPM-19 |
19 Plex |
4020.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742054
|
R&D Systems
LXSAPM-08 |
8 Plex |
1955.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741934
|
R&D Systems
LXSAPM-20 |
20 Plex |
4040.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742164
|
R&D Systems
LXSAPM-11 |
11 Plex |
2550.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762354
|
R&D Systems
LXSAPM-07 |
7 Plex |
1750.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18782324
|
R&D Systems
LXSAPM-01 |
1 Plex |
643.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772404
|
R&D Systems
LXSAPM-02 |
2 Plex |
724.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Porcine | |
Multiplex Cytokine Assay Kits | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems Porcine Luminex™ Discovery Assay