missing translation for 'onlineSavingsMsg'
Learn More
Potassium Acetate, ≥99.0%, ACS Reagent, Honeywell Fluka™ Product Code.: 15642340

Potassium Acetate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15642340 Shop All Honeywell Fluka Products
500 g, Poly Bottle, 500g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.50kg
500g
This item is not returnable. View return policy

Product Code. 15642340

Brand: Honeywell-Fluka 236497500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920Show MoreShow Less
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Physical Form Solid
Quantity 500 g
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Solubility Information Soluble in water, 98.2g/L at 20°C
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.57g/cu.cm at 25°C
Chemical Name or Material Potassium Acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Acetate, ≥99.0%, ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.