missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)
39.56€ - 373.62€
Chemical Identifiers
CAS | 298-14-6 |
---|---|
Molecular Formula | CHKO3 |
Molecular Weight (g/mol) | 100.11 |
MDL Number | MFCD00011402 |
InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
Synonym | potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 |
PubChem CID | 516893 |
ChEBI | CHEBI:81862 |
IUPAC Name | potassium;hydrogen carbonate |
SMILES | [K+].OC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15641150
|
Honeywell-Fluka
12602-1KG |
1 kg | Poly bottle |
39.56€
1kg |
In Stock
Log in to see stock. |
||||
15661150
|
Honeywell-Fluka
12602-6X1KG |
6 x 1 kg | Poly Bottle |
200.50€
Pack of 6 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
||||
15651150
|
Honeywell-Fluka
12602-25KG |
25 kg | Paper / Poly Bag |
373.62€
25kg |
In Stock
Log in to see stock. |
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
298-14-6 | |
100.11 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
516893 | |
potassium;hydrogen carbonate |
CHKO3 | |
MFCD00011402 | |
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
CHEBI:81862 | |
[K+].OC([O-])=O |
Specifications
298-14-6 | |
KHCO3 | |
NONH for all modes of transport | |
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
potassium;hydrogen carbonate | |
516893 | |
100.12g/mol | |
puriss. |
CHKO3 | |
MFCD00011402 | |
4535309 | |
Soluble in water | |
[K+].OC([O-])=O | |
100.11 | |
CHEBI:81862 | |
99.5 to 101% (acidimetric) | |
Potassium Bicarbonate, Meets analytical specification of Ph. Eur. BP, USP |
Safety and Handling
missing translation for 'einecsNumber' : 206-059-0