Learn More
Potassium Iodide Saturated, EU Pharmacopoeia Grade, Fisher Chemical™ Product Code.: 12900514
compliance-icons
Potassium Iodide Saturated, EU Pharmacopoeia Grade, Fisher Chemical™
Product Code. 12900514
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 12900514
Brand: Fisher Chemical RP 173 100
Edge
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1030582 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 173 100
Lot Number1025807 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 173 100
Lot Number1013899 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 173 100
Lot Number1031383 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberRP 173 100
compliance-icons
Product Identifier
  • Saturated Potassium Iodide Solution in carbon dioxide-free water European Pharmacopoeia Ref: 1070504
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide
Product Content Correction
Product Title
Potassium Iodide Saturated, EU Pharmacopoeia Grade, Fisher Chemical™ >