missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate Electrolyte Solution, SI Analytics™

Product Code. 11974709
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Potassium Nitrate Electrolyte Solution is used in the application and preparation of electrolyte solutions.

  • Mixture, consisting of the following components: Water, potassium nitrate, potassium chloride
  • Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
  • Protective gloves recommended for frequent or long-term skin contact
TRUSTED_SUSTAINABILITY

Specifications

CAS 7757-79-1
Melting Point −6°C
Boiling Point 104°C
Color Colorless
Density 1.12g/cm3
Concentration 0.001 M
Physical Form Liquid
Quantity 250 mL
Solution Type Potassium Nitrate
Formulation 2 mol/l KNO3 + 0.001 mol/l KCl solution
Molecular Formula KNO3
Odor Odorless
MDL Number MFCD00011409
Vapor Pressure 23hPa
Solubility Information Fully misible in water
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium nitrate
Molecular Weight (g/mol) 101.10
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'