Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF55plus
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand:  Memmert™ UF55plusH6
Product Code. 17599505
  • 298.00€ / Each
Only null left
Estimated Shipment: 21-07-2025
Add to basket
Description
Description
  • Accessory
  • For combination error message
Specifications
Specifications
UF55plus Oven
Socket to NAMUR NE 28
2 A
Potential-free Contact
24 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF55plus > For Use With: UF55plus