Learn More
Pour Point Standard IP15, ASTM D97, ISO 3016, BS 2000, Pt 15 (nominal value: -30.3 °C) UKAS ISO 17034 Certified 250mL, Paragon Scientific™
Pour Point Certified Reference Materials for the verification of laboratory instruments used in testing petroleum and derivative products for Pour Point to IP 15, ASTM D97, NFT 60-105.
Brand:  Paragon Scientific CRMU-PPLU1
Product Code. 16010060
  • 238.00€ / 250mL
Estimated Shipment: 21-02-2025
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
  • Certifications
    Nominal values quoted
Nominal values quoted. Certified Values will vary from batch to batch.