missing translation for 'onlineSavingsMsg'
Learn More

Marigold™ G07B+ Blue Nitrile Single Use Gloves

Product Code. 12989994
Click to view available options
Size:
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Flexible and comfortable glove with good dexterity.

  • Industrial protective glove
  • Sensitive yet durable
  • Blue synthetic unsupported glove
  • Lightweight unlined nitrile glove
  • Raised diamond grip pattern on palm and fingers
  • Suitable glove for all types of food
  • Excellent resistance to animal fats

Applications:

  • Food preparation, processing and handling
  • Meat and poultry processing
  • Fish processing
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Nitrile
Size 2X-Large
Thickness (Metric) 0.48 mm
Glove Cuff Style Straight
Model 9837
Length (Metric) 330 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'