Learn More
Thermo Scientific™ Precise™ 8 to 16%, Tris-HEPES, Mini, Protein Gel, 15-well
Separation Range: 14 to 205 kd; For Use With: Mini Gel Tank
Brand: Thermo Scientific™ 25243
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Precise Tris-HEPES Gels are precast polyacrylamide gels for protein electrophoresis (PAGE) that use Tris-HEPES running buffer, have long shelf lives, and provide compatibility with different commercial gel tanks.
Precise Gels are cast in a durable plastic cassette using a neutral-pH buffer that prevents polyacrylamide breakdown and results in a long shelf life. High-resolution staining and transfer of proteins is accomplished quickly on these 1mm thick mini gels. Gels are individually packaged in an easy-to-open plastic pouch and are ready to run with no comb or tape to remove. Following electrophoresis, the cassettes are easy to dismantle (no special tools required) without damaging the gel. Precise Gels are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.Highlights:
High-performance – unique running buffer produces excellent separation and high-resolution protein bands that stain easily using coomassie and silver stains
Long shelf life – guaranteed 12-month stability ensures consistent performance for the duration of each package of gels
Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes in just 90 minutes
Large-volume wells – innovative well design accommodates twice the sample volume of typical mini-gel cassette wells (10-well=50μL, 12-well=30μL; 15-well=25μL)
Compatible – cassettes fit many popular mini-gel tanks, and gels can be loaded using standard Laemmli sample loading buffers
Several convenient styles – choose from five gel densities and three well formats
Additional Features:
- Cassette dimensions: 10cm × 8.5cm × 4.5mm
- Gel dimensions: 8cm × 5.8cm × 1mm
- Storage conditions: 1 year at 4°C
- Stacking gel: 4% polyacrylamide
- Buffer system in gel: Tris-HCl, pH 7
- SDS content: none
- Required running buffer: Tris-HEPES-SDS (Part No. 28398)
- Recommended sample buffer: Tris-HCl-SDS (any type)
Specifications
8-16% Tris-HEPES Gels, 10 x 8.5cm, 15-Well | |
8 to 16% | |
4.5 mm | |
Gradient Page | |
14 to 205 kDa | |
Precast 8 to 16% gradient polyacrylamide gels for SDS-PAGE | |
15 samples per gel, 25uL capacity per well | |
100 mm |
Mini Gel Tank | |
Mini | |
Tris-HEPES | |
Protein | |
15-well | |
10/Pk. | |
Standard | |
85 mm |