Learn More
Propargyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 15264904
compliance-icons compliance-icons compliance-icons
Propargyl chloride, 98%, Thermo Scientific Chemicals
Product Code. 15264904
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Product Code. 15264904
Brand: Thermo Scientific Acros 451840250
Request Bulk or Custom Format
CAS: 624-65-7 | C3H3Cl | 74.51 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 624-65-7
Molecular Formula C3H3Cl
Molecular Weight (g/mol) 74.51
MDL Number MFCD00000980
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyneShow MoreShow Less
PubChem CID 12221
IUPAC Name 3-chloroprop-1-yne
SMILES ClCC#C
Specifications
Density 1.0300g/mL
Boiling Point 58.0°C to 61.0°C
Flash Point 19°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2Cl
Quantity 25 g
Merck Index 15,7924
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble.
Formula Weight 74.51
Percent Purity 98%
Chemical Name or Material Propargyl chloride
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466356 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number451840250,451840050
Lot NumberA0420559 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number451840250,451840050
Lot NumberA0466356 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number451840250,451840050
Lot NumberA0420559 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number451840250,451840050
Lot NumberA0394421 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number451840250,451840050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Propargyl chloride, 98%, Thermo Scientific Chemicals >