Learn More
Thermo Scientific™ ProSpecT™ Entamoeba histolytica Microplate Assay
Rapidly detect Entamoeba histolytica specific antigen from human fecal samples.
1040.00€
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | ProSpecT Entamoeba histolytica Microplate Assay |
CE Marker | Yes |
Detectable Analytes | E. histolytica Antigen |
Includes | Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
13296079
|
Thermo Scientific™
R2456096 |
ProSpecT Entamoeba histolytica Microplate Assay | 96 Tests/Kit | 2°C to 8°C |
1040.00€
96 tests |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
Description
Rapidly detect Entamoeba histolytica specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Entamoeba histolytica Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Entamoeba histolytica. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
ProSpecT Entamoeba histolytica is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Entamoeba histolytica provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
ProSpecT Entamoeba histolytica Microplate Assay | |
E. histolytica Antigen |