Learn More
Novus Biologicals™ Prostate Tissue Slides (Adult Normal)- Paraffin
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Brand: Novus Biologicals™ NBP2-67986
This item is not returnable.
View return policy
Description
The Human Prostate Tissue Lysate (Adult Normal) is derived from human.Specifications
Brain | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Fas Receptor/TNFRSF6/CD95 | |
Store at -80°C. Avoid freeze-thaw cycles. |
5 Slides | |
Normal | |
Human | |
Adult |