Learn More
Laboratoires Anios Protective Cream
This rich, robust protective cream protects the skin against bacteriostatic and fungistatic aggressions due to working conditions.
Brand:  Laboratoires Anios 399756DK
Product Code. 15666135
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Protective cream against repeated attacks bacteriostatic and fungistatic skin protection
  • Composition: cetylstearic alcohol, shea butter, glycerine, agents
  • Microbiological properties: bacteriostatic and fungistatic
  • Complies with the French Pharmacopoeia Xth edition, VIII, 14
Specifications
Specifications
Tube
50 mL
Product Title
Laboratoires Anios Protective Cream > Volume: 50mL