missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Protein Disulfide Isomerase/P4HB Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

437.00€ - 1080.00€

Specifications

Assay Range 47 pg/mL
Assay Sensitivity 47-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18250347
View Documents
Novus Biologicals™
NBP2-79305-15PLATES
15 Plates
1080.00€
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
18634660
View Documents
Novus Biologicals™
NBP2-79305-5Plates
5 Plates
437.00€
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Protein Disulfide Isomerase/P4HB.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Protein Disulfide Isomerase/P4HB Antibody Pair [HRP]

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.