missing translation for 'onlineSavingsMsg'
Learn More

PSG1 293T Cell Overexpression Lysate (Denatured), Abnova

Brand:  Abnova H00005669-T01.100uL


Product Code. 16179731

  • 215.00€ / 100µL
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Western Blotting

Specifications

Specifications

293T
PSG1 293T Cell Transient Overexpression Lysate(Denatured)
43.23
pCMV-PSG1 full-length
100 μL
Human
Western Blot
BC022338
6.8
Transient overexpression cell Lysate was tested with Anti-PSG1 antibody (H00005669-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL PSG1 293T Cell Transient Overexpression Lysa

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers