Learn More
Epredia™ PTFE Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
Brand: Epredia™ X2XER202W
128.83 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Binds and keeps liquids applied to the well area
- PTFE Construction
- Provides wettable wells for staining and growing cell cultures
- Prevents cross-contamination
- Hydrophobic coatings are very durable and withstand the strongest stains
- Autoclavable
Specifications
25 mm | |
PTFE | |
12 | |
90°, ground edge | |
100/Pk. |
75 mm | |
Diagnostic Slide | |
White | |
5.2 mm Well |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13