Learn More
BRAND™ PTFE Dropping Bottle
Click to view available options
Capacity:
25 mL
50 mL
Description
- Flexible
- Outstanding chemical resistance
- Dropper nozzle with screw cap
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 25 mL |
Includes | Dropper Nozzle |
Material | PTFE |
Closure Included | Yes |
Product Type | Dropping Bottle |
Closure Type | Screw Cap |