Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, bent
For safe connection of hoses.
Brand:  DWK Life Sciences 286230605
95.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10795295
  • 108.00€ / Pack of 10
Estimated Shipment: 12-12-2024
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Bent
9 mm
GL 14