Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, straight
For safe connection of hoses.
Brand: DWK Life Sciences 286230408
153.33 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
- Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
10/Pk. | |
Straight | |
13 mm | |
GL 25 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
17Hours
10Minutes
32Seconds
37