Learn More
Fisherbrand™ PTFE Containers
Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.
22.25€ - 311.00€
Specifications
Color | White |
---|---|
For Use With (Application) | Storage and Transport of Hazardous Chemicals |
Includes | Closure |
Material | PTFE |
Max. Temperature (Metric) | 280°C |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10313502
|
Fisherbrand™
038.015 |
15 mL |
22.25€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10323502
|
Fisherbrand™
038.030 |
30 mL |
29.05€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10140342
|
Fisherbrand™
038.060 |
60 mL |
35.35€
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
10291952
|
Fisherbrand™
038.120 |
120 mL |
48.75€
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
10199901
|
Fisherbrand™
038.240 |
240 mL |
62.20€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10724143
|
Fisherbrand™
038.360 |
360 mL |
89.30€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10393882
|
Fisherbrand™
038.480 |
480 mL |
101.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10363602
|
Fisherbrand™
038.1000 |
1000 mL |
163.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10660473
|
Fisherbrand™
038.2200 |
2200 mL |
311.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
Description
Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.
- Smooth internal finish
- Contains crew cap
- Stackable
- Autoclavable
Specifications
White | |
Closure | |
280°C | |
No | |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
Autoclavable | |
Screw Cap |
Storage and Transport of Hazardous Chemicals | |
PTFE | |
-200 to +280°C | |
No | |
Yes | |
No |