Learn More
Thermo Scientific™ PTFE One-Piece HPLC Column Connector
Ensure leak-free column connections and optimize liquid chromatography performance with the inert and biocompatible Thermo Scientific™ PTFE One-Piece HPLC Column Connector.
Brand: Thermo Scientific™ 60170-370
100.50 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Minimize HPLC system dead volume and ensure optimal separation efficiencies with the Thermo Scientific™ PTFE One-Piece HPLC Column Connector. Ideal for coupling analytical and guard columns with 10–32 threads, it provides fingertight, leak-free connections. The material is inert and biocompatible for a wide range of applications.
Specifications
Coupler | |
One-piece column coupler | |
PEEK | |
Analytical and Guard Columns with 10 to 32 Threads |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
28Hours
0Minutes
5Seconds
44 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
59Hours
0Minutes
5Seconds
44New Offer Save Up to 70%