Learn More
Fisherbrand™ PTFE Plain Stirrer Shaft
Fisherbrand™ PTFE Plain Stirrer Shafts have a PTFE encapulated stainless steel core with exposed end. They are a fully molded construction featuring a PTFE roto molded to a fully PTFE encapsulated steel reinforced drive shaft.
31.85€ - 167.00€
Specifications
Description PTFE Plain Stirrer Shaft
Quantity 1
View More Specs
Products 11
Product Code Brand Dimensions (L x W) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Diameter (Metric) Price Quantity & Availability  
16265891
View Documents Promotion Details
Fisherbrand™
005.100830EX
300 x 8 mm 5 mm
32.45€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
16275891
View Documents Promotion Details
Fisherbrand™
005.100850EX
500 x 8 mm 5 mm
38.45€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16285891
View Documents Promotion Details
Fisherbrand™
005.101030EX
300 x 10 mm 6.35 mm
31.85€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16295891
View Documents Promotion Details
Fisherbrand™
005.101050EX
500 x 10 mm 6.35 mm
38.40€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
16205901
View Documents Promotion Details
Fisherbrand™
005.101250EX
500 x 12 mm 6.35 mm
67.65€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16215901
View Documents Promotion Details
Fisherbrand™
005.101265EX
650 x 12 mm 6.35 mm
72.15€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16225901
View Documents Promotion Details
Fisherbrand™
005.101275EX
750 x 12 mm 6.35 mm
72.90€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16235901
View Documents Promotion Details
Fisherbrand™
005.1016100EX
1000 x 16 mm 10 mm
117.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16245901
View Documents Promotion Details
Fisherbrand™
005.1016750EX
750 x 16 mm 10 mm
109.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16255901
View Documents Promotion Details
Fisherbrand™
005.1019100EX
1000 x 19 mm 10 mm
167.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
16265901
View Documents Promotion Details
Fisherbrand™
005.1019750EX
750 x 19 mm 10 mm
144.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
  • PTFE with Steel Core
  • 50mm Exposed End
  • Chemically Inert
  • Easy to clean
  • No cross contamination
  • Non scratch
  • Use up to 280° C
Specifications
Specifications
PTFE Plain Stirrer Shaft
1