Learn More
Thermo Scientific™ I-Chem Septa for 24-414 Open Top Caps
missing translation for 'orderingAttributeHoverText'
Material:
PTFE-lined Silicone, Silicone
Description
- PTFE-lined silicone septum
- 0.125 in. thick or 0.060in. thin natural PTFE-lined silicone septum, 24mm diameter
- For use with Thermo Scientific VOA vials and bottles with 24-414 open-top cap
- Round shape, Natural color
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Material | PTFE-lined Silicone, Silicone |
No. per Pack | 72Pack |
Packaging | 72Pack |
Quantity | 72Pack |
Product Type | Septa |
Product Content Correction
Product Title
Thermo Scientific™ I-Chem Septa for 24-414 Open Top Caps >