Learn More
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
Brand:  Fisherbrand™ 001.315
Product Code. 16285801
  • 9.80€ / Pack of 10
Estimated Shipment: 13-06-2025
Description
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action.
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
4.5 mm
10/Pk.
PTFE Stirrer Bars Pivot Ring
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
PTFE
15 mm
Cylinder, Pivot ring
Product Title
Fisherbrand™ PTFE Stir Bars with Pivot Ring >