Learn More
RETSCH Pure 70mm Diameter Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200
Brand:  RETSCH 03.647.0290
 View more versions of this product
Product Code. 11721856
  • 216.00€ / Each
Estimated Shipment: 03-02-2025
Specifications
1Each
Ring Sieve
Ultra Centrifugal Mill ZM 200
0.25 mm
Stainless Steel
70 mm