Learn More
Thermo Scientific™ Purified Water for Microbiology
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Purified Water for Microbiology.
Brand: Thermo Scientific™ BO0184B
Description
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Thermo Scientific™ Oxoid™ Purified Water. The bottles come with screw cap closure for ease of operation.
Use Purified Water as a diluting and rinsing fluid.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Colorless, clear | |
Sterile Distilled Water | |
United Kingdom | |
3.0g + 0.3g |
2 to 25°C | |
Tubes with screw cap closure | |
Purified Water | |
300 x 3 mL in Tube |