Learn More
Karl Hecht™ Pycnometers According to Gay-Lussac
For the measurement of liquid density in the laboratory.
74.20€ - 81.00€
Specifications
Type | Pycnometer acc. to Gay-Lussac |
---|---|
For Use With (Application) | Measuring liquid density |
Description | With ground-in stopper |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10758102
|
Karl Hecht™
42571008 |
5 mL |
74.20€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
10217341
|
Karl Hecht™
42571032 |
50 mL |
81.00€
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
Description
- With ground-in stopper
- Precisely adjusted
- Available in different capacities
Specifications
Pycnometer acc. to Gay-Lussac | |
Measuring liquid density | |
With ground-in stopper |
Product Title
Karl Hecht™ Pycnometers According to Gay-Lussac