Learn More
Pyrex™ Gas Distribution Tube
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
Brand:  Pyrex 3830/02M
70.58 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 12663232
  • 82.40€ / Pack of 2
Only null left
Estimated Shipment: 18-08-2025
Description
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
  • Ideal for the dispersion of gas in liquids
  • Borosilicate glass tube with sintered thimble
  • Choice of thimble porosities to achieve optimum gas bubble size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Porosity: 1
300 mm
Sintered thimble
Borosilicate Glass (3.3)
For the dispersion of gas in liquids
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Pyrex™ Gas Distribution Tube > Porosity: 1