Learn More
Pyrex™ Polytetrafluoroethylene Spare Keys
Brand:  Pyrex SST108K
Product Code. 15728067
  • 127.00€ / Each
Estimated Shipment: 16-05-2025
Specifications
Stopcock Key
8 mm
PTFE
Product Title
Pyrex™ Polytetrafluoroethylene Spare Keys > Plug Bore: 8mm