missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Q-VD-OPH Inhibitor Peptide
Applications: Functional, In vitro assay
396.00€ - 1340.00€
Specifications
Color | Off-white |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748003
|
Novus Biologicals™
NBP2-29391 |
396.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
18110683
|
Novus Biologicals™
NBP2-29391-3MG |
3 mg |
975.00€
3mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18180784
|
Novus Biologicals™
NBP2-29391-5MG |
5 mg |
1340.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Off-white | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Q-VD-OPH Inhibitor Peptide