missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qa-1(b) Mouse anti-Mouse , PE, Clone: 6A8.6F10.1A6, BD Biosciences™
Mouse Monoclonal Antibody
318.00€ - 780.00€
Specifications
Antigen | Qa-1(b) |
---|---|
Clone | 6A8.6F10.1A6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17207514
|
BD Biosciences
566640 |
25 μg |
318.00€
25µg |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
17217514
|
BD Biosciences
566641 |
0.1 mg |
780.00€
0.10mg |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
Description
Qa-1(b) Monoclonal antibody specifically detects Qa-1(b) in Mouse samples. It is validated for Flow CytometrySpecifications
Qa-1(b) | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Qa-1[b] aa. 161-179 Peptide | |
Primary |
6A8.6F10.1A6 | |
Monoclonal | |
Mouse | |
Mouse | |
H2-Qa1b; H2-T23; Qa-1(b); Qa-1b; Qa1b; T23b | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |