missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Quantofix™ Potassium Semi-quantitative Test Strips
Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
Brand: Macherey-Nagel™ 91316
Additional Details : Weight : 0.24000kg
Description
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
Yellow to Orange | |
Determination of Potassium | |
100 Pack | |
1 min. |
Strip | |
0, 200, 400, 700, 1000, 1500mg/L K+ | |
30°C | |
Semi-qualitative Test Strip |