Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System, AB Assurance extended warranty, SmartStart, 384-well
The Applied Biosystems QuantStudio 7™ Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Package includes the QuantStudio 7™ Pro Real-Time PCR System, SmartSmart™ Orientation, and a one-year extended warranty.
Brand: Applied Biosystems™ A45583
This item is not returnable.
View return policy
Description
- Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint
- The QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
- Designed for ultimate ease-of-use, the system offers high quality, high throughput, and excellent reliability
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- An optimal user experience for researchers who want to work smarter in the lab
- Package includes the QuantStudio 7™ Pro Real-Time PCR System, SmartSmart™ Orientation, and a one-year extended warranty
- Warranty includes comprehensive service coverage and one planned maintenance vist
Services
1 year extended
Specifications
Interchangeable | |
Real Time Thermal Cyclers | |
12 in. Color Touchscreen | |
Room Temperature | |
1 package |
QuantStudio™ 7 Pro Real Time PCR System (384-well, 0.02 mL block), 1 year extended Warranty, and SmartStart™ orientation | |
QuantStudio™ | |
Energy Efficient, Fewer Resources used and Less Waste | |
QuantStudio 7 Pro |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System, AB Assurance extended warranty, SmartStart, 384-well > Room Temperature