Learn More
Hellma™ Quartz Suprasil Ultramicro Cell
Brand: Hellma™ 1052541540
This item is not returnable.
View return policy
Specifications
Ultra-micro Cell | |
Quartz Suprasil | |
12.5 mm |
45 μL | |
45 mm |
Product Content Correction
Product Title
Hellma™ Quartz Suprasil Ultramicro Cell > Capacity: 45μL