Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFit™ Borosilicate glass drying angled cone to fit QuickFitT™ apparatus.
Brand: Quickfit MF23/3
41.75 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- For use with a desiccant such as calcium chloride
- Ground glass cone to fit Quickfit™ apparatus
Specifications
Quickfit™ apparatus | |
Borosilicate Glass | |
Tube | |
24/29 |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14