Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA21
29.31 EUR valid until 2025-10-26
Use promo code "24136" to get your promotional price.
 View more versions of this product
Product Code. 12146220
  • ONLINE EXCLUSIVE PRICE
    39.35€
    37.98€ / Pack of 2
    Save 1.37€
    3% Off
View Available Promotions
Only null left
Estimated Shipment: 30-10-2025
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/23
Borosilicate Glass
19/26
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 14/23; Socket size: 19/26