missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Polyacetal Clamp for Spherical Joints

For retaining glassware having Quickfit™ spherical joints

Brand:  Quickfit KS35

46.03 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 12623032

  • 48.45€ / Pack of 5
Estimated Shipment: 31-07-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from polyacetal
  • Can considerably reduce the need for clamping
  • Easy to fit and remove
  • Can be used at temperatures up to 80°C
  • Clips are color coded for easy identification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5/Pk.
Joint Clamp
Polyacetal
Light Orange
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quickfit™ Polyacetal Clamp for Spherical Joints > Joint Size: S35; Color: Light orange

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.