Learn More
Quickfit™ Silicone Rings for Screw Thread Glass Joints
For use with Quickfit™ glass screw threads
5.30€ - 77.70€
Product Code | Brand | Diameter (Metric) Hole | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Hole | Size | Price | Quantity & Availability | ||||
12919283
|
Quickfit
QR28/11 |
10.5 to 11.5 mm | Thread size: 28mm |
77.70€
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
12969273
|
Quickfit
QR18/7 |
7 to 8.5 mm | Thread size: 18mm |
23.60€
Pack of 10 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
12959273
|
Quickfit
QR13/6 |
6 to 7mm | Thread size: 13mm |
16.10€
Pack of 10 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
12999273
|
Quickfit
QR24/11 |
10.5 to 11.5 mm | Thread size: 24mm |
41.30€
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
12979273
|
Quickfit
QR24/6 |
6 to 7mm | Thread size: 24mm |
40.40€
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
12989273
|
Quickfit
QR24/9 |
8.5 to 9.5mm | Thread size: 24mm | N/A | |||||
11392075
|
Quickfit
QR28/18 |
18.0 to 19.0mm | Thread size: 28mm | N/A | |||||
11302085
|
Quickfit
QR28/6 |
6 to 7mm | Thread size: 28mm |
67.45€
Pack of 10 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||
15187193
|
Quickfit
QR28/6 |
6 to 7mm | Thread size: 28mm | N/A | |||||
Description
For use with Quickfit™ glass screwthreads
- For use with Quickfit™ glass screwthreads
- Melamine screwcaps with aperture at the top
- Silicone rubber rings with PTFE washers
Specifications
Silicone |