Learn More
Contec™ Quiltec™ I Mop Head
Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
Brand: Contec™ MHQT0100
135.43 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Two-ply 100% knitted polyester over a rayon core
- Gamma irradiated at no less than 25 kGy
- 5 mops double bagged
- Approx 18-20 sqm coverage
- Certificates of conformity supplied with every batch
Specifications
Mop Head | |
Non-sterile | |
39 x 13 cm | |
5/Bag, 8 Bags/Cs. | |
Quilted Polyester |
White | |
Not Autoclavable | |
Single-use | |
No |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
022377 | Certificate of Quality | 31/01/2025 | MHQT0100 |
021857 | Certificate of Quality | 08/10/2024 | MHQT0100 |
021482 | Certificate of Quality | 16/08/2024 | MHQT0100 |
020971 | Certificate of Quality | 16/05/2024 | MHQT0100 |
1052784-0-0-1 | Certificate of Compliance | 06/08/2024 | MHQT0100 |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02