missing translation for 'onlineSavingsMsg'
Learn More
Quinoxaline, 98+%, Thermo Scientific Chemicals Product Code.: 10508893

Quinoxaline, 98+%, Thermo Scientific Chemicals

Product Code. 10508893
500 g, Glass Bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10508893

Brand: Thermo Scientific Acros 132225000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-19-0
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006719
InChI Key XSCHRSMBECNVNS-UHFFFAOYSA-N
Synonym benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalinShow MoreShow Less
PubChem CID 7045
ChEBI CHEBI:36616
IUPAC Name quinoxaline
SMILES C1=CC=C2C(=C1)N=CC=N2

Specifications

Melting Point 29°C to 34°C
Color Yellow to Brown
Boiling Point 220°C to 223°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 500 g
Beilstein 23, 176
Merck Index 15, 8195
Solubility Information Solubility in water: soluble. Other solubilities: miscible in alcohol,ether,benzene,acetone
Formula Weight 130.15
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Quinoxaline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0392436 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number132225000,132221000
Lot NumberA0392022 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number132225000,132221000
Lot NumberA0426493 Certificate TypeCertificate of Analysis Date01/02/2021 Catalog Number132225000,132221000
Lot NumberA0421967 Certificate TypeCertificate of Analysis Date01/09/2020 Catalog Number
Product Identifier
  • Quinoxaline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quinoxaline, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.