missing translation for 'onlineSavingsMsg'
Learn More
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid Product Code.: 11492230

(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Product Code. 11492230
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11492230

Brand: Thermo Scientific Alfa Aesar H29112.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103733-65-9 | C10H11NO2 | 177.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103733-65-9
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00144038
InChI Key BWKMGYQJPOAASG-UHFFFAOYNA-N
Synonym r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
PubChem CID 712398
SMILES OC(=O)C1CC2=CC=CC=C2CN1

Specifications

Quantity 5 g
Optical Rotation +163° (c=2 in 1N NaOH)
IUPAC Name (3R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Formula Weight 177.2
Chemical Name or Material (R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10141595 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29112.06,H29112.MD,H29112.03
Lot Number10235672 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29112.06,H29112.MD,H29112.03
Lot Number61500177 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29112.06,H29112.MD,H29112.03
Lot Number10232040 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29112.06,H29112.MD,H29112.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.