missing translation for 'onlineSavingsMsg'
Learn More
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals Product Code.: 15495445

(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 15495445
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15495445

Brand: Thermo Scientific Alfa Aesar H60112.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Incompatible with light and oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-98-1
Molecular Formula C76H108O8P2
Molecular Weight (g/mol) 1211.64
MDL Number MFCD20265636
InChI Key KACYLFSRRUJDSY-UHFFFAOYSA-N
Synonym r-dtbm-garphos, 4,4',6,6'-tetramethoxy-1,1'-biphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, ar-2,2'-bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino-4,4'-dimethoxy-6,6'-dimethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, s-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, 2'-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 3,5-di-tert-butyl-4-methoxyphenyl phosphaneShow MoreShow Less
PubChem CID 87110675
IUPAC Name [2-[2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphaneShow MoreShow Less
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC(=C(C(=C2)C(C)(C)C)OC)C(C)(C)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C(C)(C)C)OC)C(C)(C)C)C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)OC)OC)OC)OCShow MoreShow Less

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 1211.61
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW21E043 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberH60112.03,H60112.MD
Lot NumberP14F041 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberH60112.03,H60112.MD
Lot NumberZ15D026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60112.03,H60112.MD
Lot NumberP14F041 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60112.03,H60112.MD
Lot NumberW21E043 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60112.03,H60112.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.