Learn More
R&D Systems™ Human CD43 (NP_003114) VersaClone cDNA
Cluster of Differentiation 43
Brand:  R&D Systems™ RDC0071
Product Code. 18461760
  • 322.00€ / 10µg
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Human CD43 (NP_003114) VersaClone cDNA clone contains the complete ORF for CD43/Sialophorin, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD43/Sialophorin insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hCD43
10 μg at 0.2 μg/μL
1216bp
Human
NP_003114
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans