Learn More
R&D Systems™ Human EMR3 (NP_115960) VersaClone cDNA
EGF-like Module Containing, Mucin-like, Hormone Receptor-like 3
Brand: R&D Systems™ RDC0686
This item is not returnable.
View return policy
Description
The Human EMR3 (NP_115960) VersaClone cDNA clone contains the complete ORF for EMR3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The EMR3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hEMR3 | |
10 ÎĽg at 0.2 ÎĽg/ÎĽL | |
1972bp | |
Human |
NP_115960 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 ÎĽg |
Product Content Correction
Product Title
R&D Systems™ Human EMR3 (NP_115960) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans